missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-(2-hydroxyethyl)perfluorooctylsulphonamide (Technical Grade), TRC

Product Code. 30136697
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136697 10 g 10g
30137626 5 g 5g
30040961 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136697 Supplier Toronto Research Chemicals Supplier No. TRCE91765010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1691-99-2
Molecular Formula C12 H10 F17 N O3 S
Molecular Weight (g/mol) 571.25
Synonym N-Ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(2-hydroxyethyl)-1-octanesulfonamide, 2-(N-Ethylperfluoro-1-octanesulfonamido)ethanol, 2-Perfluorooctylsulfonyl-N-ethylaminoethyl alcohol, 2-[N-Ethyl-N-(perfluorooctylsulfonyl)amino]ethanol, AI 3-29782, FC 10, Fluorad FC 10, LE 011, N-EtFOSE
IUPAC Name N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(2-hydroxyethyl)octane-1-sulfonamide
SMILES CCN(CCO)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H10F17NO3S/c1-2-30(3-4-31)34(32,33)12(28,29)10(23,24)8(19,20)6(15,16)5(13,14)7(17,18)9(21,22)11(25,26)27/h31H,2-4H2,1H3
Formula Weight 571.011
Chemical Name or Material N-Ethyl-N-(2-hydroxyethyl)perfluorooctylsulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.