missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-(2-hydroxyethyl)perfluorooctanesulfonamide Phosphate Diester, TRC

Product Code. 30378287
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378287 25 mg
30376301 250 mg
30384876 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378287 Supplier Toronto Research Chemicals Supplier No. TRCE67883525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2965-52-8
Molecular Formula C24 H19 F34 N2 O8 P S2
Molecular Weight (g/mol) 1204.47
Synonym 1-Octanesulfonamide, N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(2-hydroxyethyl)-, phosphate (ester) (2:1) (8CI), N,N'-[Phosphinicobis(oxy-2,1-ethanediyl)]bis[N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-octanesulfonamide], N-Ethyl-N-(2-hydroxyethyl)perfluorooctanesulfonamide phosphate diester, diSAmPAP, 1-Octanesulfonamide, N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(2-hydroxyethyl)-, phosphate (ester) (2:1) (8CI), N,N'-[Phosphinicobis(oxy-2,1-ethanediyl)]bis[N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-octanesulfonamide], N-Ethyl-N-(2-hydroxyethyl)perfluorooctanesulfonamide phosphate diester
IUPAC Name bis[2-[ethyl(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctylsulfonyl)amino]ethyl] hydrogen phosphate
SMILES CCN(CCOP(=O)(O)OCCN(CC)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H19F34N2O8PS2/c1-3-59(70(63,64)23(55,56)19(45,46)15(37,38)11(29,30)9(25,26)13(33,34)17(41,42)21(49,50)51)5-7-67-69(61,62)68-8-6-60(4-2)71(65,66)24(57,58)20(47,48)16(39,40)12(31,32)10(27,28)14(35,36)18(43,44)22(52,53)54/h3-8H2,1-2H3,(H,61,62)
Formula Weight 1203.9778
Chemical Name or Material Bis(2-perfluorooctylsulfonyl-N-ethylaminoethyl) phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.