missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-(2-hydroxyethyl)nitrosamine, TRC

Product Code. 30381919
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381919 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381919 Supplier Toronto Research Chemicals Supplier No. TRCE678860100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13147-25-6
Molecular Formula C4H10N2O2
Quantity 100 mg
Synonym 2-(Ethylnitrosoamino)ethanol, N-(2-Hydroxyethyl)-N-ethylnitrosamine, N-Nitroso-2-(ethylamino)ethanol, N-Nitroso-N-ethyl-2-hydroxyethylamine, N-Nitrosoethyl-2-hydroxyethylamine
InChI Formula InChI=1S/C4H10N2O2/c1-2-6(5-8)3-4-7/h7H,2-4H2,1H3
SMILES CCN(CCO)N=O
Molecular Weight (g/mol) 118.13
Formula Weight 118.0742
Chemical Name or Material N-Ethyl-N-(2-hydroxyethyl)nitrosamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.