missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl Maleimide, TRC

Product Code. 30370245
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30370245 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370245 Supplier Toronto Research Chemicals Supplier No. TRCE8179001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Ethyl Maleimide
CAS 128-53-0
Molecular Formula C6 H7 N O2
Quantity 1 g
Synonym 1H-Pyrrole-2,5-dione, 1-ethyl- (9CI, ACI), 1-Ethyl-1H-pyrrole-2,5-dione (ACI), Maleimide, N-ethyl- (8CI), 1-Ethyl-2,5-dihydro-1H-pyrrole-2,5-dione, Ethylmaleimide, Maleic acid N-ethylimide, N-Ethylmaleimide, NEM, NSC 7638, N-Ethyl maleimide
InChI Formula InChI=1S/C6H7NO2/c1-2-7-5(8)3-4-6(7)9/h3-4H,2H2,1H3
SMILES CCN1C(=O)C=CC1=O
IUPAC Name 1-ethylpyrrole-2,5-dione
Molecular Weight (g/mol) 125.13
Formula Weight 125.0477
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.