missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl Maleimide (1M Solution in Acetonitrile), TRC

Product Code. 30407453
Change view
Click to view available options
Quantity:
1x5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407453 1x5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407453 Supplier Toronto Research Chemicals Supplier No. TRCKIT41591X5ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Ethyl Maleimide
CAS 128-53-0
Molecular Formula C6 H7 N O2
Quantity 1x5 mL
Synonym 1H-Pyrrole-2,5-dione, 1-ethyl- (9CI, ACI), 1-Ethyl-1H-pyrrole-2,5-dione (ACI), Maleimide, N-ethyl- (8CI), 1-Ethyl-2,5-dihydro-1H-pyrrole-2,5-dione, Ethylmaleimide, Maleic acid N-ethylimide, N-Ethylmaleimide, NEM, NSC 7638, N-Ethyl maleimide
InChI Formula InChI=1S/C6H7NO2/c1-2-7-5(8)3-4-6(7)9/h3-4H,2H2,1H3
SMILES CCN1C(=O)C=CC1=O
IUPAC Name 1-ethylpyrrole-2,5-dione
Molecular Weight (g/mol) 125.13
Formula Weight 125.0477
Concentration or Composition (by Analyte or Components) 1M
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.