missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-D-glucamine, TRC

Product Code. 30387566
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387566 50 g 50g
30375826 100 g -
30385661 250 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387566 Supplier Toronto Research Chemicals Supplier No. TRCE67894550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14216-22-9
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.24
Synonym 1-Deoxy-1-(ethylamino)-D-glucitol, D-(-)-N-Ethylglucamine, Eglumine, N-Ethylglucamine, (2R,3R,4R,5S)-6-(Ethylamino)hexane-1,2,3,4,5-pentaol
IUPAC Name (2R,3R,4R,5S)-6-(ethylamino)hexane-1,2,3,4,5-pentol
SMILES CCNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

Specifications

Quantity 50 g
InChI Formula InChI=1S/C8H19NO5/c1-2-9-3-5(11)7(13)8(14)6(12)4-10/h5-14H,2-4H2,1H3/t5-,6+,7+,8+/m0/s1
Formula Weight 209.1263
Chemical Name or Material N-Ethyl-D-glucamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.