missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.

Product Code. 10071920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071920 100 g Glass bottle 100g
10685792 25 g Glass bottle 25g
10512354 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10071920 Supplier Thermo Scientific Acros Supplier No. 166031000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7570-45-8 | C15H13NO | 223.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7570-45-8
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.28
MDL Number MFCD00004963
InChI Key QGJXVBICNCIWEL-UHFFFAOYSA-N
Synonym 9-ethyl-9h-carbazole-3-carbaldehyde, n-ethyl-3-carbazolecarboxaldehyde, n-ethylcarbazole-3-carboxaldehyde, 3-formyl-n-ethylcarbazole, n-ethylcarbazole-3-carbaldehyde, 9-ethyl-3-carbazolecarboxaldehyde, ccris 9052, n-ethyl carbazole-3-aldehyde, n-ethyl-carbazole-3-aldehyde, pubchem9556
PubChem CID 82055
IUPAC Name 9-ethylcarbazole-3-carbaldehyde
SMILES CCN1C2=C(C=CC=C2)C2=C1C=CC(C=O)=C2

Tekniske data

CAS Min % 92.0
CAS Max % 100.0
Melting Point 85°C to 91°C
Color Yellow to Ochre
Infrared Spectrum Authentic
Assay Percent Range 92% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 223.27
Percent Purity 94%
Grade Technical
Physical Form Crystalline Powder or Granules
Chemical Name or Material N-Ethyl-3-carbazolecarboxaldehyde
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.