missing translation for 'onlineSavingsMsg'
Learn More

N-[Ethyl-(2-pyridyl-5-ethyl) Pioglitazone(Pioglitazone Impurity), TRC

Product Code. 30388728
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388728 50 mg
30373572 100 mg
30385290 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388728 Supplier Toronto Research Chemicals Supplier No. TRCE92575050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 952188-00-0
Molecular Formula C28 H31 N3 O3 S
Molecular Weight (g/mol) 489.63
Synonym 2,4-Thiazolidinedione, 5-[[4-[2-(5-ethyl-2-pyridinyl)ethoxy]phenyl]methyl]-3-[2-(5-ethyl-2-pyridinyl)ethyl]-, 5-[[4-[2-(5-Ethyl-2-pyridinyl)ethoxy]phenyl]methyl]-3-[2-(5-ethyl-2-pyridinyl)ethyl]-2,4-thiazolidinedione, 5-[4-[2-(5-Ethylpyridin-2-yl)ethoxy]benzyl]-3-[2-(5-ethylpyridin-2-yl)ethyl]thiazolidine-2,4-dione, (5RS)-5-[4-[2-(5-Ethylpyridin-2-yl)ethoxy]benzyl]-3-[2-(5-ethylpyridin-2-yl)ethyl]-1,3-thiazolidine-2,4-dione, (5RS)-5-[[4-[2-(5-Ethylpyridin-2-yl)ethoxy]phenyl]methyl]-3-[2-(5-ethylpyridin-2-yl)ethyl]thiazolidine-2,4-dione
IUPAC Name 5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-3-[2-(5-ethylpyridin-2-yl)ethyl]-1,3-thiazolidine-2,4-dione
SMILES CCc1ccc(CCOc2ccc(CC3SC(=O)N(CCc4ccc(CC)cn4)C3=O)cc2)nc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C28H31N3O3S/c1-3-20-5-9-23(29-18-20)13-15-31-27(32)26(35-28(31)33)17-22-7-11-25(12-8-22)34-16-14-24-10-6-21(4-2)19-30-24/h5-12,18-19,26H,3-4,13-17H2,1-2H3
Formula Weight 489.2086
Chemical Name or Material (5RS)-5-[[4-[2-(5-Ethylpyridin-2-yl)ethoxy]phenyl]methyl]-3-[2-(5-ethylpyridin-2-yl)ethyl]thiazolidine-2,4-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.