missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-2-nitroaniline, 98%, Thermo Scientific™

Product Code. p-7052646
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15451308

Brand: Thermo Scientific Alfa Aesar H55148.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10112-15-9
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00007091
InChI Key CQIKVOWCSGXCCG-UHFFFAOYSA-N
Synonym benzenamine,n-ethyl-2-nitro, benzenamine, n-ethyl-2-nitro, n-ethyl-2-nitroaniline, ethyl-2-nitro-phenyl-amine, ethyl-6-nitroaniline, acmc-20ak4v, ethyl 2-nitrophenyl amine, n-ethyl-n-2-nitrophenyl amine #
PubChem CID 82354
IUPAC Name N-ethyl-2-nitroaniline
SMILES CCNC1=CC=CC=C1[N+](=O)[O-]

Specifications

Color Colorless to Orange
Density 1.19 g/mL
Boiling Point 168°C (20 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.645
Quantity 250 mg
Formula Weight 166.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Ethyl-2-nitroaniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.