missing translation for 'onlineSavingsMsg'
Learn More

N-(Ethoxycarbonyl)phthalimide, 97%

Product Code. 15433097
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433097 25 g 25g
15443097 100 g 100g
15453097 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15433097 Supplier Thermo Scientific Alfa Aesar Supplier No. A14822.14
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 22509-74-6 | C11H9NO4 | 219.196 g/mol

N-(Ethoxycarbonyl)phthalimide is used to protect amine functional groups. Reagent for the conversion of amines and amino acids to their phthaloyl derivatives under mild conditions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(Ethoxycarbonyl)phthalimide is used to protect amine functional groups. Reagent for the conversion of amines and amino acids to their phthaloyl derivatives under mild conditions

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22509-74-6
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.196
MDL Number MFCD00005893
InChI Key VRHAQNTWKSVEEC-UHFFFAOYSA-N
Synonym n-carbethoxyphthalimide, n-ethoxycarbonylphthalimide, n-carboethoxyphthalimide, phthalimide-n-carbethoxy, ethyl n-phthaloylcarbamate, n-ethoxycarbonyl phthalimide, 2h-isoindole-2-carboxylic acid, 1,3-dihydro-1,3-dioxo-, ethyl ester, ethyl 1,3-dioxoisoindoline-2-carboxylate, n-karbetoksi-ftalimid, 2-isoindolinecarboxylic acid, 1,3-dioxo-, ethyl ester
PubChem CID 31187
IUPAC Name ethyl 1,3-dioxoisoindole-2-carboxylate
SMILES CCOC(=O)N1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 78°C to 84°C
Quantity 25 g
Beilstein 196340
Solubility Information Insoluble in water.
Formula Weight 219.2
Percent Purity 97%
Chemical Name or Material N-(Ethoxycarbonyl)phthalimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.