missing translation for 'onlineSavingsMsg'
Learn More

N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, Thermo Scientific Chemicals

Catalog Number p-3736425
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16357-59-8
C14H17NO3
247.29
MFCD00006703
GKQLYSROISKDLL-UHFFFAOYNA-N
eedq, 2-ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, n-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, ethyl 2-ethoxyquinoline-1 2h-carboxylate, 1-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 2-ethoxy-1 2h-quinolinecarboxylic acid, ethyl ester, 1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester, ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate, ethyl 1,2-dihydro-2-ethoxy-1-quinolinecarboxylate, ethyl n-2-ethoxy-1,2-dihydroquinoline carboxylate
27833
ethyl 2-ethoxy-2H-quinoline-1-carboxylate
CCOC1C=CC2=CC=CC=C2N1C(=O)OCC
This item is not returnable. View return policy
16357-59-8
C14H17NO3
247.29
MFCD00006703
GKQLYSROISKDLL-UHFFFAOYNA-N
eedq, 2-ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, n-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, ethyl 2-ethoxyquinoline-1 2h-carboxylate, 1-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 2-ethoxy-1 2h-quinolinecarboxylic acid, ethyl ester, 1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester, ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate, ethyl 1,2-dihydro-2-ethoxy-1-quinolinecarboxylate, ethyl n-2-ethoxy-1,2-dihydroquinoline carboxylate
27833
ethyl 2-ethoxy-2H-quinoline-1-carboxylate
CCOC1C=CC2=CC=CC=C2N1C(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 63.0°C to 66.0°C
Color White to Beige
Boiling Point 125.0°C to 128.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 21, V,3, 28
Fieser 02,191; 03,137; 04,223
Merck Index 15, 3566
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 247.29
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material N-Ethoxycarbonyl-2-ethoxy-1, 2-dihydroquinoline
Show More Show Less
compliance-icons
Product Identifier
  • N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'