missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(epsilon)-Acetyl-L-lysine, 99%

Product Code. 15466219
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466219 5 g 5g
15456219 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466219 Supplier Thermo Scientific Chemicals Supplier No. J64139.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 692-04-6 | C8H16N2O3 | 188.23 g/mol

N(epsilon)-Acetyl-L-lysine is an R-chain N-acetylated α amino acid used together with other lysine analogues to differentiate and characterized various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N(epsilon)-Acetyl-L-lysine is an R-chain N-acetylated α amino acid used together with other lysine analogues to differentiate and characterized various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins.

Solubility
Soluble in water (miscible), and 80% acetic acid (50 mg/ml).

Notes
Hygroscopic. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 692-04-6
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.23
MDL Number MFCD00002639
InChI Key DTERQYGMUDWYAZ-ZETCQYMHSA-N
PubChem CID 92832
ChEBI CHEBI:17752
IUPAC Name (2S)-6-acetamido-2-aminohexanoic acid
SMILES CC(=O)NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 250°C (decomposition)
Color White
Synonym n-epsilon-acetyl-l-lysine, n6-acetyl-l-lysine, nepsilon-acetyl-l-lysine, n-epsilon-acetyllysine, h-lys ac-oh, n6-acetyllysine, s-6-acetamido-2-aminohexanoic acid, n 6-acetyl-l-lysine, n 6-acetyllysine, n epsilon-acetyl-l-lysine
Solubility Information Soluble in water (miscible),and 80% acetic acid (50mg/ml).
Quantity 5 g
Formula Weight 188.2
Sensitivity Hygroscopic
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N(epsilon)-Acetyl-L-lysine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.