Learn More
n-Eicosane, 99%, Thermo Scientific Chemicals
£19.10 - £41.80
Chemical Identifiers
CAS | 112-95-8 |
---|---|
Molecular Formula | C20H42 |
Molecular Weight (g/mol) | 282.56 |
MDL Number | MFCD00009344 |
InChI Key | CBFCDTFDPHXCNY-UHFFFAOYSA-N |
Synonym | eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane |
PubChem CID | 8222 |
ChEBI | CHEBI:43619 |
IUPAC Name | icosane |
SMILES | CCCCCCCCCCCCCCCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11421518
|
Thermo Scientific Alfa Aesar
A13853.14 |
25 g |
£19.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11431518
|
Thermo Scientific Alfa Aesar
A13853.22 |
100 g |
£41.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
n-Eicosane is a saturated aliphatic hydrocarbon, which is used to form candles and paraffin waxes with solar energy storage capacity. It is also used in cosmetics, lubricants, plasticizers and in the petrochemical industry. Further, it is useful for special surfactants and in the preparation of certain organic chemicals. In addition to this, it is used as a chromatographic analysis reference material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Eicosane is a saturated aliphatic hydrocarbon, which is used to form candles and paraffin waxes with solar energy storage capacity. It is also used in cosmetics, lubricants, plasticizers and in the petrochemical industry. Further, it is useful for special surfactants and in the preparation of certain organic chemicals. In addition to this, it is used as a chromatographic analysis reference material.
Solubility
Soluble in ether, petroleum ether and benzene. Slightly soluble in acetone and chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
112-95-8 | |
282.56 | |
CBFCDTFDPHXCNY-UHFFFAOYSA-N | |
8222 | |
icosane |
C20H42 | |
MFCD00009344 | |
eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane | |
CHEBI:43619 | |
CCCCCCCCCCCCCCCCCCCC |
Specifications
112-95-8 | |
0.7888 | |
176°C (348°F) | |
C20H42 | |
MFCD00009344 | |
eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane | |
CBFCDTFDPHXCNY-UHFFFAOYSA-N | |
icosane | |
8222 | |
282.54 | |
n-Eicosane |
36°C to 38°C | |
220°C (30 mm) | |
99% | |
1.4425 | |
1700722 | |
Soluble in ether,petroleum ether and benzene. Slightly soluble in acetone and chloroform. Insoluble in water. | |
CCCCCCCCCCCCCCCCCCCC | |
282.56 | |
CHEBI:43619 | |
99% |
Safety and Handling
P301+P310-P331-P501c
H304
missing translation for 'einecsNumber' : 204-018-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only