missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyltrimethoxysilane, 95%

Product Code. 11339195
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339195 25 g 25g
11349195 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339195 Supplier Thermo Scientific Alfa Aesar Supplier No. L16486.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3069-21-4 | C15H34O3Si | 290.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3069-21-4
Molecular Formula C15H34O3Si
Molecular Weight (g/mol) 290.52
MDL Number MFCD00069148
InChI Key SCPWMSBAGXEGPW-UHFFFAOYSA-N
Synonym n-dodecyltrimethoxysilane, silane, dodecyltrimethoxy, lauryltrimethoxysilane, dodecyl trimethoxy silane, n-dodecytrimethoxysilane, acmc-1cr18, ksc494k8f
PubChem CID 76479
IUPAC Name dodecyl(trimethoxy)silane
SMILES CCCCCCCCCCCC[Si](OC)(OC)OC

Especificaciones

Boiling Point 125°C (1.8mmHg)
Melting Point -40°C
Quantity 25 g
Beilstein 2356323
Flash Point 108°C (226°F)
Formula Weight 290.52
Percent Purity 95%
Refractive Index 1.4274
Sensitivity Moisture sensitive
Density 0.89
Chemical Name or Material n-Dodecyltrimethoxysilane
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.