missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecylpyridinium chloride hydrate, 98%

Product Code. 11435497
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435497 25 g 25g
11445497 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435497 Supplier Thermo Scientific Alfa Aesar Supplier No. B23492.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207234-02-4 | C17H30ClN | 283.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207234-02-4
Molecular Formula C17H30ClN
Molecular Weight (g/mol) 283.88
MDL Number MFCD00011987
InChI Key GKQHIYSTBXDYNQ-UHFFFAOYSA-M
Synonym 1-dodecylpyridin-1-ium chloride hydrate, laurylpyridinum chloride, 1-dodecylpyridinium chloride hydrate, 1-dodecylpyridin-1-ium hydrate chloride, c17h30n.cl.h2o, n-dodecylpyridinium chloride hydrate, dodecylpyridinium chloride monohydrate, n-dodecylpyridinium chloride monohydrate, 1-dodecylpyridin-1-ium chloride-water 1/1/1
PubChem CID 16211802
IUPAC Name 1-dodecylpyridin-1-ium;chloride;hydrate
SMILES [Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1

Specifications

Melting Point 88°C to 90°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 283.89 (Anhydrous)
Percent Purity 98%
Chemical Name or Material n-Dodecylpyridinium chloride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.