missing translation for 'onlineSavingsMsg'
Learn More

N-Dodecylmethylamine, 98%

Product Code. 11357096
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357096 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11357096 Supplier Thermo Scientific Alfa Aesar Supplier No. L07025.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7311-30-0 | C13H29N | 199.382 g/mol

N-Dodecylmethylamine is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Dodecylmethylamine is used as intermediates.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7311-30-0
Molecular Formula C13H29N
Molecular Weight (g/mol) 199.382
MDL Number MFCD00026574
InChI Key OMEMQVZNTDHENJ-UHFFFAOYSA-N
Synonym n-methyldodecylamine, dodecyl methyl amine, dodecylmethylamine, 1-dodecanamine, n-methyl, n-dodecylmethylamine, dodecyl-methyl-amine, unii-88nn43i94s, methyldodecylamine, n-methyldodecanamine, n-methyl-n-dodecylamine
PubChem CID 81746
IUPAC Name N-methyldodecan-1-amine
SMILES CCCCCCCCCCCCNC

Specifications

Density 0.795
Boiling Point 90°C to 92°C (0.9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4395
Quantity 5 g
UN Number UN2735
Beilstein 1739959
Solubility Information Not miscible in water.
Formula Weight 199.38
Percent Purity 98%
Chemical Name or Material N-Dodecylmethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.