missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyl methyl sulfide, Thermo Scientific™

Product Code. 11397088
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397088 25 g 25g
11307098 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397088 Supplier Thermo Scientific Alfa Aesar Supplier No. 014877.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3698-89-3
Molecular Formula C13H28S
Molecular Weight (g/mol) 216.43
MDL Number MFCD00015073
InChI Key KJWHJDGMOQJLGF-UHFFFAOYSA-N
Synonym dodecyl methyl sulfide, 2-thiatetradecane, 1-methylsulfanyl dodecane, methylthiododecane, methyl lauryl sulfide, dodecane, 1-methylthio, methyl n-dodecyl sulfide, dodecyl methyl sulphide, sulfide, dodecyl methyl, unii-77bo1dv08u
PubChem CID 77287
IUPAC Name 1-methylsulfanyldodecane
SMILES CCCCCCCCCCCCSC

Specifications

Boiling Point 163°C to 165°C (19 mmHg)
Odor Sulphurous
Linear Formula CH3(CH2)11SCH3
Refractive Index 1.4608
Quantity 25 g
Formula Weight 216.41
Physical Form Liquid
Chemical Name or Material n-Dodecyl methyl sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.