missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyl-beta-D-maltopyranoside

Product Code. 15448399
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448399 1 g 1g
15468399 500 mg 500mg
15458399 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448399 Supplier Thermo Scientific Alfa Aesar Supplier No. J66869.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A non-ionic detergent useful in membrane solubilization studies | CAS: 69227-93-6 | C24H46O11 | 510.621 g/mol

N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.

Solubility
Soluble in water at 10% w/v

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69227-93-6
Molecular Formula C24H46O11
Molecular Weight (g/mol) 510.621
MDL Number MFCD00043012
InChI Key NLEBIOOXCVAHBD-QKMCSOCLSA-N
Synonym n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside
PubChem CID 114880
ChEBI CHEBI:43769
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O

Specifications

Melting Point 224°C to 226°C
Color White
Quantity 1 g
Beilstein 55318
Solubility Information Soluble in water at 10% w/v
Formula Weight 510.62
Physical Form Powder
Chemical Name or Material n-Dodecyl-beta-D-maltopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.