missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyl beta-D-glucopyranoside, Thermo Scientific Chemicals

Product Code. 15457429
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457429 1 g 1g
15467429 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457429 Supplier Thermo Scientific Alfa Aesar Supplier No. J66173.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59122-55-3
Molecular Formula C18H36O6
Molecular Weight (g/mol) 348.48
MDL Number MFCD00063298
InChI Key PYIDGJJWBIBVIA-UYTYNIKBSA-N
Synonym dodecyl glucoside, dodecyl beta-d-glucopyranoside, dodecyl b-d-glucopyranoside, dodecyl-beta-d-glucopyranoside, dodecyl, a-d-glucopyranoside, n-dodecyl-beta-d-glucopyranoside, n-dodecyl glucoside, n-dodecyl-i 2-d-glucopyranoside, dodecyl-, n-dodecyl ?-d-glucopyranoside
PubChem CID 93321
IUPAC Name (2R,3R,4S,5S,6R)-2-dodecoxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O

Specifications

Color White
Quantity 1 g
Beilstein 86236
Solubility Information Soluble in methanol at 50mg/ml
Formula Weight 348.47
Physical Form Powder
Chemical Name or Material n-Dodecyl beta-D-glucopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.