missing translation for 'onlineSavingsMsg'
Learn More

Melford n-Dodecyl-B-D-maltoside

Product Code. 30018225
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
Unit Size:
10g
1g
25g
50g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018225 50 g 50g
30018222 10 g 10g
30018224 5 g 5g
30018223 25 g 25g
30018221 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30018225 Supplier Melford Supplier No. D1200050.0
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Non-ionic detergent used in membrane protein studies.

Non-ionic detergent used in membrane protein studies. Reported to increase thermal stability of lipid-free rhodopsin in solution.

Specifications

Chemical Name or Material n-Dodecyl-B-D-maltoside
CAS 69227-93-6
Quantity 50 g
Molecular Formula C24H46O11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.