missing translation for 'onlineSavingsMsg'
Learn More

Melford n-Dodecyl-B-D-maltoside

Product Code. 30018222
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
Unit Size:
10g
1g
25g
50g
5g
Product Code. Quantity unitSize
30018222 10 g 10g
30018224 5 g 5g
30018225 50 g 50g
30018223 25 g 25g
30018221 1 g 1g
5 options
This item is not returnable. View return policy

Product Code. 30018222

Brand: Melford D1200010.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-ionic detergent used in membrane protein studies.

Non-ionic detergent used in membrane protein studies. Reported to increase thermal stability of lipid-free rhodopsin in solution.

Specifications

Chemical Name or Material n-Dodecyl-B-D-maltoside
CAS 69227-93-6
Quantity 10 g
Molecular Formula C24H46O11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.