missing translation for 'onlineSavingsMsg'
Learn More

N-Dodecanoylglycine-d23, TRC

Product Code. 30371752
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30371752 10 mg
30380894 2.5 mg
30384110 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30371752 Supplier Toronto Research Chemicals Supplier No. TRCD49464310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C14H4D23NO3
Molecular Weight (g/mol) 280.51
Synonym Lauroylglycine-d23, N-Lauroylglycine-d23, NSC 404235-d23, N-(1-Oxododecyl)glycine-d23
IUPAC Name (dodecanoyl-d23)glycine
SMILES O=C(NCC(O)=O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H27NO3/c1-2-3-4-5-6-7-8-9-10-11-13(16)15-12-14(17)18/h2-12H2,1H3,(H,15,16)(H,17,18)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2
Formula Weight 280.34
Chemical Name or Material N-Dodecanoylglycine-d23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.