missing translation for 'onlineSavingsMsg'
Learn More

n-Docosane, 99%

Product Code. 10591364 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10591364 500 g Glass bottle 500g
10436810 25 g Glass bottle 25g
10446810 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10591364 Supplier Thermo Scientific Acros Supplier No. 158785000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 629-97-0 | C22H46 | 310.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-97-0
Molecular Formula C22H46
Molecular Weight (g/mol) 310.61
MDL Number MFCD00009348
InChI Key HOWGUJZVBDQJKV-UHFFFAOYSA-N
Synonym n-docosane, dokosan, unii-ow99q363ko, heneicosane, methyl, ch3-ch2 20-ch3, docosane, analytical standard, twt, normal-docosane, docosane, n, docosane
PubChem CID 12405
ChEBI CHEBI:46050
IUPAC Name docosane
SMILES CCCCCCCCCCCCCCCCCCCCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 43.0°C to 46.0°C
Color White
Density 0.7700g/mL
Boiling Point 369.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)20CH3
Quantity 500 g
Beilstein 01, 174
Specific Gravity 0.77
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene and toluene
Formula Weight 310.6
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material n-Docosane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.