Product Code. 10436810
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
10436810
25 g
Glass bottle
25g
10446810
100 g
Glass bottle
100g
10591364
500 g
Glass bottle
500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
10436810
Supplier
Thermo Scientific Acros
Supplier No.
158780250
21.76 GBP valid until 2026-12-31 BEST PRICE on Promo! Use promo code "28038" to get your promotional price.
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
CAS: 629-97-0 | C22H46 | 310.61 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
629-97-0
Molecular Formula
C22H46
Molecular Weight (g/mol)
310.61
MDL Number
MFCD00009348
InChI Key
HOWGUJZVBDQJKV-UHFFFAOYSA-N
Synonym
n-docosane, dokosan, unii-ow99q363ko, heneicosane, methyl, ch3-ch2 20-ch3, docosane, analytical standard, twt, normal-docosane, docosane, n, docosane
PubChem CID
12405
ChEBI
CHEBI:46050
IUPAC Name
docosane
SMILES
CCCCCCCCCCCCCCCCCCCCCC
Specifications
CAS Min %
98.5
CAS Max %
100.0
Melting Point
43.0°C to 46.0°C
Color
White
Density
0.7700g/mL
Boiling Point
369.0°C
Flash Point
>112°C
Infrared Spectrum
Authentic
Assay Percent Range
98.5% min. (GC)
Packaging
Glass bottle
Linear Formula
CH3 (CH2 )2 0 CH3
Quantity
25 g
Beilstein
01, 174
Specific Gravity
0.77
Solubility Information
Solubility in water: insoluble. Other solubilities: soluble in benzene and toluene
Formula Weight
310.6
Percent Purity
99%
Physical Form
Crystalline Solid
Chemical Name or Material
n-Docosane
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?