missing translation for 'onlineSavingsMsg'
Learn More

N-(Dithiocarboxy)sarcosine Diammonium Salt, TRC

Product Code. 30387929
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387929 500 mg 500mg
30387013 250 mg -
30371389 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387929 Supplier Toronto Research Chemicals Supplier No. TRCD494350500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29664-09-3
Molecular Formula C4 H7 N O2 S2 . 2 H3 N
Molecular Weight (g/mol) 199.29
IUPAC Name azane;2-[dithiocarboxy(methyl)amino]acetic acid
SMILES N.N.CN(CC(=O)O)C(=S)S

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H7NO2S2.2H3N/c1-5(4(8)9)2-3(6)7;;/h2H2,1H3,(H,6,7)(H,8,9);2*1H3
Formula Weight 199.0449
Chemical Name or Material N-(Dithiocarboxy)sarcosine, Diammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.