missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Diphenylmethylene)glycine t-Butyl Ester, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | N-(Diphenylmethylene)glycine, t-Butyl Ester |
| CAS | 81477-94-3 |
| Molecular Formula | C19 H21 N O2 |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C19H21NO2/c1-19(2,3)22-17(21)14-20-18(15-10-6-4-7-11-15)16-12-8-5-9-13-16/h4-13H,14H2,1-3H3 |
| SMILES | CC(C)(C)OC(=O)CN=C(c1ccccc1)c2ccccc2 |
| IUPAC Name | tert-butyl 2-(benzhydrylideneamino)acetate |
| Molecular Weight (g/mol) | 295.38 |
| Formula Weight | 295.1572 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction