missing translation for 'onlineSavingsMsg'
Learn More

N-(Diphenylmethylene)glycine ethyl ester, 98%

Product Code. 11477256
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477256 25 g 25g
11467256 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477256 Supplier Thermo Scientific Alfa Aesar Supplier No. B25580.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69555-14-2 | C17H17NO2 | 267.328 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69555-14-2
Molecular Formula C17H17NO2
Molecular Weight (g/mol) 267.328
MDL Number MFCD00010590
InChI Key QUGJYNGNUBHTNS-UHFFFAOYSA-N
Synonym ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylidene amino acetate, diphenylmethylene glycine ethylester, ethyl diphenylmethylenamino acetate, diphenylmethylene glycine ethyl ester
PubChem CID 319508
IUPAC Name ethyl 2-(benzhydrylideneamino)acetate
SMILES CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 51°C to 55°C
Quantity 25 g
Flash Point >110°C (230°F)
Formula Weight 267.33
Sensitivity Moisture sensitive
Percent Purity 98%
Chemical Name or Material N-(Diphenylmethylene)glycine ethyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.