missing translation for 'onlineSavingsMsg'
Learn More

N-(Diphenylmethylene)aminoacetonitrile, 99%

Product Code. 10214840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10214840 5 g Glass bottle 5g
10637484 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10214840 Supplier Thermo Scientific Acros Supplier No. 124140050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70591-20-7 | C15H12N2 | 220.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70591-20-7
Molecular Formula C15H12N2
Molecular Weight (g/mol) 220.28
MDL Number MFCD00009970
InChI Key VRLJFRODHVSTIK-UHFFFAOYSA-N
Synonym n-diphenylmethylene aminoacetonitrile, 2-diphenylmethylene amino acetonitrile, benzhydrylidene-amino-acetonitrile, 2-benzhydrylideneamino acetonitrile, 2-diphenylmethylidene amino acetonitrile, acetonitrile, diphenylmethylene amino, 4,4-diphenyl-3-azabut-3-enenitrile, pubchem13883, benzhydrylideneaminoacetonitrile
PubChem CID 612957
IUPAC Name 2-(benzhydrylideneamino)acetonitrile
SMILES N#CCN=C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 83.0°C to 86.0°C
Color Gray or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)2C=NCH2CN
Quantity 5 g
Formula Weight 220.27
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material N-(Diphenylmethylene)aminoacetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.