missing translation for 'onlineSavingsMsg'
Learn More

Globus Shetland™ Showa Best Gloves™ N-DEX Free™ Disposable Green Nitrile Gloves

Product Code. 12981075
Click to view available options
Size:
7
8
9
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12981075

Brand: Globus Shetland™ BSTI77052

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Wear Showa Best Gloves™ N-DEX Free™ Disposable Green Nitrile Gloves for high visibility. The patented, accelerator-free formulation makes these gloves a PPE Category III item.

  • Free from natural rubber latex and Type I allergic reactions
  • Textured fingertips
  • Rolled cuff
  • Green
  • Powder-free

Specifications

Certifications/Compliance CE, EN374-2:1994, EN374-3:1994, CATEGORY III
Color Green
Disposable Single-use
Exterior Finish Partially Textured
Laboratory/Cleanroom Use Yes
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 8
Thickness (Metric) 0.10 mm
Glove Cuff Style Rolled
Model 7705PFT-08
Series 7705PFT
Length (Metric) 220 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.