missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyltramadol Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30457907
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457907 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457907 Supplier LoGiCal Supplier No. LGCAMP0007.0701
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

N-Desmethyltramadol Hydrochloride 1.0 mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Desmethyl-cis-tramadol Hydrochloride
CAS 1018989-94-0
Analyte or Component Names N-Desmethyl-cis-tramadol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C15 H23 N O2 . Cl H
Certifications/Compliance ISO 17034
Synonym N-Desmethyltramadol Hydrochloride, Cyclohexanol, 1-(3-methoxyphenyl)-2-[(methylamino)methyl]-, hydrochloride (1:1), (1 R,2 R)-rel-, N-Demethyltramadol hydrochloride
InChI Formula InChI=1 S/C15H23NO2.ClH/c1-16-11-13-6-3-4-9-15(13,17)12-7-5-8-14(10-12)18-2;/h5,7-8,10,13,16-17 H,3-4,6,9,11H2,1-2H3;1 H/t13-,15+;/m1./s1
SMILES Cl.CNC[C@H]1 CCCC[C@]1(O)c2cccc(OC)c2
IUPAC Name (1 R,2 R)-1-(3-methoxyphenyl)-2-(methylaminomethyl)cyclohexan-1-ol;hydrochloride
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 285.81
Quantity 1 mL
Formula Weight 285.1496 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.