missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyldiltiazem Hydrochloride, LoGiCal

Product Code. 30460256
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460256 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460256 Supplier LoGiCal Supplier No. LGCFOR0068.03
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Desmethyldiltiazem Hydrochloride
CAS 130606-60-9
Analyte or Component Names N-Desmethyldiltiazem Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C21 H24 N2 O4 S . Cl H
Certifications/Compliance ISO 9001
Synonym Diltiazem Hydrochloride Imp. D (EP), Diltiazem Imp. D (EP), (2S,3S)-2-(4-Methoxyphenyl)-5-[2-(methylamino)ethyl]-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl Acetate Hydrochloride, Diltiazem Hydrochloride Imp. D (EP) as Hydrochloride, N-Desmethyl-diltiazem Hydrochloride, Diltiazem Hydrochloride Impurity D as Hydrochloride, Diltiazem Impurity D as Hydrochloride
InChI Formula InChI=1S/C21H24N2O4S.ClH/c1-14(24)27-19-20(15-8-10-16(26-3)11-9-15)28-18-7-5-4-6-17(18)23(21(19)25)13-12-22-2;/h4-11,19-20,22H,12-13H2,1-3H3;1H/t19-,20+;/m1./s1
Solution Type Neat
SMILES Cl.CNCCN1C(=O)[C@H](OC(=O)C)[C@@H](Sc2ccccc12)c3ccc(OC)cc3
IUPAC Name [(2S,3S)-2-(4-methoxyphenyl)-5-[2-(methylamino)ethyl]-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 436.95
Quantity 10 mg
Formula Weight 436.1224 g/mol
Show More Show Less
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.