missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethylcitalopram.HCl, 1mg/ml in Methanol (as free base), Lipomed

Product Code. 30461175
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461175 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461175 Supplier Lipomed Supplier No. LPMCTL1888HC1LM
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

N-Desmethylcitalopram.HCl, 1mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmethylcitalopram Hydrochloride
CAS 97743-99-2
Analyte or Component Names Desmethylcitalopram Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H19 F N2 O . Cl H
Concentration 1 mg/mL
Synonym Citalopram Hydrobromide Imp. D (EP), Citalopram Imp. D (EP), Citalopram USP Related Compound D, Citalopram USP RC D, (1 RS)-1-(4-Fluorophenyl)-1-[3-(methylamino)propyl]-1,3-dihydroisobenzofuran-5-carbonitrile Hydrochloride, Lu 11-109 C, Lu 11-109 C, Lu 11-109 C, Lu 11-109 C, Lu 11-109 C, Lu 11-109 C, Lu 11-109 C, Lu 11-109 C, Lu 11-109 C
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.CNCCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3
IUPAC Name 1-(4-fluorophenyl)-1-[3-(methylamino)propyl]-3 H-2-benzofuran-5-carbonitrile;hydrochloride
Solvent or Matrix Methanol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 346.83
Quantity 1 mL
Formula Weight 346.1248 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.