missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Nefopam-d5 Hydrochloride (>90%), TRC

Product Code. 30377457
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377457 10 mg -
30378207 2.5 mg 2.5mg
30385212 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30377457 Supplier Toronto Research Chemicals Supplier No. TRCD42235710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

Molecular Formula C16H12D5NO • HCl
Molecular Weight (g/mol) 280.8
Synonym 3,4,5,6-Tetrahydro-1-phenyl-1H-2,5-benzoxazocine-D5 Hydrochloride, N-Desmethylnefopam-D5 Hydrochloride, Nornefopam-D5 Hydrochloride
IUPAC Name 1-(phenyl-d5)-3,4,5,6-tetrahydro-1H-benzo[f][1,4]oxazocine hydrogen chloride
SMILES [2H]C1=C(C([2H])=C(C([2H])=C1[2H])[2H])C2OCCNCC3=CC=CC=C32.Cl

Specifikationer

Quantity 10 mg
InChI Formula InChI=1S/C16H17NO.ClH/c1-2-6-13(7-3-1)16-15-9-5-4-8-14(15)12-17-10-11-18-16;/h1-9,16-17H,10-12H2;1H/i1D,2D,3D,6D,7D;
Formula Weight 280.14
Chemical Name or Material N-Desmethyl Nefopam-D5 Hydrochloride
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.