missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Diltiazem Hydrochloride, TRC

Product Code. 30134042
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134042 50 mg 50mg
17973715 5 mg 5mg
17933265 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134042 Supplier Toronto Research Chemicals Supplier No. TRCD29161050MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130606-60-9
Molecular Formula C21 H24 N2 O4 S . Cl H
Molecular Weight (g/mol) 436.95
Synonym (2S,3S)-2-(4-Methoxyphenyl)-5-[2-(methylamino)ethyl]-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl Acetate Hydrochloride, Diltiazem Hydrochloride Imp. D (EP) as Hydrochloride, N-Desmethyl-diltiazem Hydrochloride
IUPAC Name [(2S,3S)-2-(4-methoxyphenyl)-5-[2-(methylamino)ethyl]-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride
SMILES Cl.CNCCN1C(=O)[C@H](OC(=O)C)[C@@H](Sc2ccccc12)c3ccc(OC)cc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H24N2O4S.ClH/c1-14(24)27-19-20(15-8-10-16(26-3)11-9-15)28-18-7-5-4-6-17(18)23(21(19)25)13-12-22-2;/h4-11,19-20,22H,12-13H2,1-3H3;1H/t19-,20+;/m1./s1
Formula Weight 436.1224
Chemical Name or Material N-Desmethyldiltiazem Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.