missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Dextrorphan Hydrobromide, TRC

Product Code. 30136966
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136966 25 mg 25mg
30006879 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136966 Supplier Toronto Research Chemicals Supplier No. TRCD29159025MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1058187-40-8
Molecular Formula C16 H21 N O . Br H
Molecular Weight (g/mol) 324.26
Synonym Morphinan-3-ol, hydrobromide (1:1), (9α,13α,14α)-, (+)-3-Hydroxymorphinan hydrobromide, Nordextrorphan Hydrobromide
SMILES Br.Oc1ccc2C[C@@H]3NCC[C@]4(CCCC[C@H]34)c2c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H21NO.BrH/c18-12-5-4-11-9-15-13-3-1-2-6-16(13,7-8-17-15)14(11)10-12;/h4-5,10,13,15,17-18H,1-3,6-9H2;1H/t13-,15+,16+;/m1./s1
Formula Weight 323.0885
Chemical Name or Material N-Desmethyl Dextrorphan Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.