missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Danofloxacin, TRC

Product Code. 30005579
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005579 1 mg 1mg
30003318 5 mg 5mg
30007690 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005579 Supplier Toronto Research Chemicals Supplier No. TRCD2915851MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108461-04-7
Molecular Formula C18 H18 F N3 O3
Molecular Weight (g/mol) 343.35
IUPAC Name 1-cyclopropyl-7-[(1S,4S)-2,5-diazabicyclo[2.2.1]heptan-2-yl]-6-fluoro-4-oxoquinoline-3-carboxylic acid
SMILES OC(=O)C1=CN(C2CC2)c3cc(N4C[C@@H]5C[C@H]4CN5)c(F)cc3C1=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H18FN3O3/c19-14-4-12-15(5-16(14)21-7-9-3-11(21)6-20-9)22(10-1-2-10)8-13(17(12)23)18(24)25/h4-5,8-11,20H,1-3,6-7H2,(H,24,25)/t9-,11-/m0/s1
Formula Weight 343.1332
Chemical Name or Material N-Desmethyldanofloxacin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.