missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Danofloxacin, TRC

Product Code. 30362690
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362690 50 mg 50mg
30354032 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30362690 Supplier Toronto Research Chemicals Supplier No. TRCD29158550MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Desmethyldanofloxacin
CAS 108461-04-7
Molecular Formula C18 H18 F N3 O3
Quantity 50 mg
InChI Formula InChI=1S/C18H18FN3O3/c19-14-4-12-15(5-16(14)21-7-9-3-11(21)6-20-9)22(10-1-2-10)8-13(17(12)23)18(24)25/h4-5,8-11,20H,1-3,6-7H2,(H,24,25)/t9-,11-/m0/s1
SMILES OC(=O)C1=CN(C2CC2)c3cc(N4C[C@@H]5C[C@H]4CN5)c(F)cc3C1=O
IUPAC Name 1-cyclopropyl-7-[(1S,4S)-2,5-diazabicyclo[2.2.1]heptan-2-yl]-6-fluoro-4-oxoquinoline-3-carboxylic acid
Molecular Weight (g/mol) 343.35
Formula Weight 343.1332
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.