missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Clozapine, TRC

Product Code. 17914385
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17914385 25 mg 25mg
17970935 5 mg 5mg
17993115 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17914385 Supplier Toronto Research Chemicals Supplier No. TRCD29200025MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6104-71-8
Molecular Formula C17 H17 Cl N4
Molecular Weight (g/mol) 312.8
Synonym 5H-Dibenzo[b,e][1,4]diazepine, 8-chloro-11-(1-piperazinyl)-, 8-Chloro-11-(piperazin-1-yl)-5H-dibenzo[b,e][1,4]diazepine, Clozapine Imp. C (EP), Desmethylclozapine, Norclozapine, 8-Chloro-11-(1-piperazinyl)-5H-dibenzo[b,e][1,4]diazepine, ACP 104, Demethylclozapine, N-Demethylclozapine, N-Desmethylclozapine, Clozapine-desmethyl
IUPAC Name 3-chloro-6-piperazin-1-yl-11H-benzo[b][1,4]benzodiazepine
SMILES Clc1ccc2Nc3ccccc3C(=Nc2c1)N4CCNCC4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H17ClN4/c18-12-5-6-15-16(11-12)21-17(22-9-7-19-8-10-22)13-3-1-2-4-14(13)20-15/h1-6,11,19-20H,7-10H2
Formula Weight 312.1142
Percent Purity >95%
Chemical Name or Material N-Desmethylclozapine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.