missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl-benzotropine Hydrochloride, TRC

Product Code. 30364293
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364293 250 mg 250mg
30363993 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30364293 Supplier Toronto Research Chemicals Supplier No. TRCD291040250MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 25471-67-4
Molecular Formula C20 H23 N O . Cl H
Molecular Weight (g/mol) 329.86
Synonym 8-Azabicyclo[3.2.1]octane, 3-(diphenylmethoxy)-, hydrochloride, endo- (9CI), 1αH,5αH-Nortropane, 3α-(diphenylmethoxy)-, hydrochloride (8CI), (3-endo)-3-(Diphenylmethoxy)-8-azabicyclo[3.2.1]octane hydrochloride, endo-3-(Benzhydryloxy)-8-azabicyclo[3.2.1]octane hydrochloride, Desmethyl benzatropine hydrochloride, N-Desmethylbenzatropine hydrochloride, N-Desmethylbenztropine hydrochloride
IUPAC Name (1R,5S)-3-benzhydryloxy-8-azabicyclo[3.2.1]octane;hydrochloride
SMILES Cl.C1C[C@H]2C[C@H](C[C@@H]1N2)OC(c3ccccc3)c4ccccc4

Tekniske data

Quantity 250 mg
InChI Formula InChI=1S/C20H23NO.ClH/c1-3-7-15(8-4-1)20(16-9-5-2-6-10-16)22-19-13-17-11-12-18(14-19)21-17;/h1-10,17-21H,11-14H2;1H/t17-,18+,19+;
Formula Weight 329.1546
Chemical Name or Material N-Desmethyl Benzotropine hydrochloride
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.