missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Azatadine, TRC

Product Code. 17953465
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
10mg
2.5mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17953465 50 mg 50mg
17953165 10 mg 10mg
17953275 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17953465 Supplier Toronto Research Chemicals Supplier No. TRCD29094050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38092-95-4
Molecular Formula C19 H20 N2
Molecular Weight (g/mol) 276.3755
Synonym 6,11-Dihydro-11-(4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
IUPAC Name 11-piperidin-4-ylidene-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
SMILES C1CC(=C2c3ccccc3CCc4cccnc24)CCN1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H20N2/c1-2-6-17-14(4-1)7-8-16-5-3-11-21-19(16)18(17)15-9-12-20-13-10-15/h1-6,11,20H,7-10,12-13H2
Formula Weight 276.1626
Chemical Name or Material N-Desmethyl Azatadine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.