missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl-6-O-methylerythromycin (9E)-Oxime, TRC

Product Code. 30135446
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135446 50 mg 50mg
30007590 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135446 Supplier Toronto Research Chemicals Supplier No. TRCD29231050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127182-43-8
Molecular Formula C37 H68 N2 O13
Molecular Weight (g/mol) 748.94
Synonym 3' '-N-Demethyl-6-O-methylerythromycin A (E)-9-oxime, Oxacyclotetradecane, erythromycin deriv., Clarithromycin Imp. M (EP)
IUPAC Name (3R,4S,5S,6R,7R,9R,10E,11S,12R,13S,14R)-14-ethyl-12,13-dihydroxy-10-hydroxyimino-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-6-[(2S,3R,4S,6R)-3-hydroxy-6-methyl-4-(methylamino)oxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecan-2-one
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@H](NC)[C@H]3O)[C@@](C)(C[C@@H](C)\C(=N/O)\[C@H](C)[C@@H](O)[C@]1(C)O)OC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C37H68N2O13/c1-14-25-37(10,44)30(41)20(4)27(39-45)18(2)16-36(9,47-13)32(52-34-28(40)24(38-11)15-19(3)48-34)21(5)29(22(6)33(43)50-25)51-26-17-35(8,46-12)31(42)23(7)49-26/h18-26,28-32,34,38,40-42,44-45H,14-17H2,1-13H3/b39-27+/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1
Formula Weight 748.4721
Chemical Name or Material N-Desmethyl-6-O-methylerythromycin A (9E)-Oxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.