missing translation for 'onlineSavingsMsg'
Learn More

N-Desisopropyl Metoprolol, TRC

Product Code. 30363275
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363275 1 g 1g
30352033 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30363275 Supplier Toronto Research Chemicals Supplier No. TRCD2902401G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74027-60-4
Molecular Formula C12 H19 N O3
Molecular Weight (g/mol) 225.28
Synonym 2-Propanol, 1-amino-3-[4-(2-methoxyethyl)phenoxy]-, N-Desisopropylmetoprolol
IUPAC Name 1-amino-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol
SMILES COCCc1ccc(OCC(O)CN)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H19NO3/c1-15-7-6-10-2-4-12(5-3-10)16-9-11(14)8-13/h2-5,11,14H,6-9,13H2,1H3
Formula Weight 225.1365
Chemical Name or Material N-Desisopropylmetoprolol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.