missing translation for 'onlineSavingsMsg'
Learn More

N-Desethyl Milnacipran-d5, TRC

Product Code. 30357907
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357907 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30357907 Supplier Toronto Research Chemicals Supplier No. TRCD28940210MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Desethyl Milnacipran-d5
CAS 1217609-30-7
Molecular Formula C13 H13 D5 N2 O
Quantity 10 mg
Synonym N-Desethyl Milnacipran D5
InChI Formula InChI=1S/C13H18N2O/c1-2-15-12(16)13(8-11(13)9-14)10-6-4-3-5-7-10/h3-7,11H,2,8-9,14H2,1H3,(H,15,16)/t11-,13+/m1/s1/i1D3,2D2
SMILES [2H]C([2H])([2H])C([2H])([2H])N=C(O)[C@@]1(C[C@@H]1CN)c2ccccc2
Molecular Weight (g/mol) 223.33
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.