missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-des-succinyl-N-Boc-O-desethyl Sacubitril, TRC
Specifications
Specifications
| Chemical Name or Material | (2R,4S)-5-(Biphenyl-4-yl)-4-[(tert-butoxycarbonyl)amino]-2-methylpentanoic Acid |
| CAS | 1012341-50-2 |
| Molecular Formula | C23H29NO4 |
| Quantity | 250 mg |
| Synonym | (2R,4S)-5-(Biphenyl-4-yl)-4-[(tert-butoxycarbonyl)amino]-2-methylpentanoic Acid, (alphaR,gammaS)-gamma-[[(1,1-Dimethylethoxy)carbonyl]amino]-alpha-methyl[1,1'-biphenyl]-4-pentanoic Acid |
| InChI Formula | InChI=1S/C23H29NO4/c1-16(21(25)26)14-20(24-22(27)28-23(2,3)4)15-17-10-12-19(13-11-17)18-8-6-5-7-9-18/h5-13,16,20H,14-15H2,1-4H3,(H,24,27)(H,25,26)/t16-,20+/m1/s1 |
| SMILES | C[C@H](C[C@@H](Cc1ccc(cc1)c2ccccc2)NC(=O)OC(C)(C)C)C(=O)O |
| IUPAC Name | (2R,4S)-2-methyl-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(4-phenylphenyl)pentanoic acid |
| Molecular Weight (g/mol) | 383.48 |
| Formula Weight | 383.2097 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Ser du en mulighed for forbedring?