missing translation for 'onlineSavingsMsg'
Learn More

N-Demethyl Trimipramine Hydrochloride, TRC

Product Code. 30349939
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349939 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349939 Supplier Toronto Research Chemicals Supplier No. TRCD2345601MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmethyltrimipramine Hydrochloride
CAS 10464-24-1
Molecular Formula C19 H24 N2 . Cl H
Quantity 1 mg
Synonym (2RS)-3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,2-dimethylpropan-1-amine Hydrochloride, Trimipramine Maleate Imp. B (EP) as Hydrochloride, Trimipramine-desmethyl Hydrochloride
InChI Formula InChI=1S/C19H24N2.ClH/c1-15(13-20-2)14-21-18-9-5-3-7-16(18)11-12-17-8-4-6-10-19(17)21;/h3-10,15,20H,11-14H2,1-2H3;1H
SMILES Cl.CNCC(C)CN1c2ccccc2CCc3ccccc13
IUPAC Name 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,2-dimethylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 316.87
Formula Weight 316.1706
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.