missing translation for 'onlineSavingsMsg'
Learn More

N-Demethyl Rifampicin, TRC

Product Code. 30366478
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366478 1 mg 1mg
30363025 10 mg 10mg
30357967 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366478 Supplier Toronto Research Chemicals Supplier No. TRCD2312501MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-45-0
Molecular Formula C42 H56 N4 O12
Molecular Weight (g/mol) 808.91
Synonym N-Demethylrifampicin, Rifamycin, 3-[(1-piperazinylimino)methyl]-, 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione, 5,6,9,17,19,21-hexahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-8-(N-1-piperazinylformimidoyl)-, 21-acetate (7CI,8CI), 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan, rifamycin deriv., 3-[(1-Piperazinylimino)methyl]rifamycin, 3-Piperazinoiminomethyl rifamycin SV, 4-N-Demethylrifampicin, AF/AP, Demethylrifampicin, N-Demethyl rifampicin, N-Desmethylrifampicin, N-Demethylrifampin, NSC 143416, Rifampicin AF/AP
SMILES CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(=O)C)[C@@H]1C)\C)c(\C=N\N5CCNCC5)c(O)c4c3C2=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C42H56N4O12/c1-20-11-10-12-21(2)41(54)45-32-27(19-44-46-16-14-43-15-17-46)36(51)29-30(37(32)52)35(50)25(6)39-31(29)40(53)42(8,58-39)56-18-13-28(55-9)22(3)38(57-26(7)47)24(5)34(49)23(4)33(20)48/h10-13,18-20,22-24,28,33-34,38,43,48-52H,14-17H2,1-9H3,(H,45,54)/b11-10+,18-13+,21-12-,44-19+/t20-,22+,23+,24+,28-,33-,34+,38+,42-/m0/s1
Formula Weight 808.3895
Chemical Name or Material N-Demethyl Rifampin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.