missing translation for 'onlineSavingsMsg'
Learn More

N-Demethyl Mifepristone, TRC

Product Code. 30004263
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004263 50 mg 50mg
30010027 10 mg 10mg
30003614 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004263 Supplier Toronto Research Chemicals Supplier No. TRCD23095050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104004-96-8
Molecular Formula C28 H33 N O2
Molecular Weight (g/mol) 415.57
Synonym Estra-4,9-dien-3-one, 17-hydroxy-11-[4-(methylamino)phenyl]-17-(1-propyn-1-yl)-, (11β,17β)-, Estra-4,9-dien-3-one, 17-hydroxy-11-[4-(methylamino)phenyl]-17-(1-propynyl)-, (11β,17β)- (9CI), (11β,17β)-17-Hydroxy-11-[4-(methylamino)phenyl]-17-(1-propyn-1-yl)estra-4,9-dien-3-one, Desmethylmifepristone, Metapristone, N-Demethylmifepristone, N-Desmethyl-RU 486, N-Methylaminopristone, RU 42633, N-Demethyl Mifepristone
IUPAC Name (8S,11R,13S,14S,17S)-17-hydroxy-13-methyl-11-[4-(methylamino)phenyl]-17-prop-1-ynyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
SMILES CNc1ccc(cc1)[C@H]2C[C@@]3(C)[C@@H](CC[C@@]3(O)C#CC)[C@@H]4CCC5=CC(=O)CCC5=C24

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C28H33NO2/c1-4-14-28(31)15-13-25-23-11-7-19-16-21(30)10-12-22(19)26(23)24(17-27(25,28)2)18-5-8-20(29-3)9-6-18/h5-6,8-9,16,23-25,29,31H,7,10-13,15,17H2,1-3H3/t23-,24+,25-,27-,28-/m0/s1
Formula Weight 415.2511
Percent Purity >95%
Chemical Name or Material N-Demethyl Mifepristone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.