missing translation for 'onlineSavingsMsg'
Learn More

N-Demethyl Ivabradine Hydrochloride, TRC

Product Code. 30356602
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356602 25 mg 25mg
30352081 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30356602 Supplier Toronto Research Chemicals Supplier No. TRCD29300025MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Demethyl Ivabradine Hydrochloride
CAS 1246638-08-3
Molecular Formula C26 H34 N2 O5 . Cl H
Quantity 25 mg
Synonym 2H-3-Benzazepin-2-one, 3-[3-[[[(7S)-3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]amino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-, hydrobromide (1:1)
InChI Formula InChI=1S/C26H34N2O5.ClH/c1-30-22-11-17-6-9-28(26(29)14-18(17)12-23(22)31-2)8-5-7-27-16-20-10-19-13-24(32-3)25(33-4)15-21(19)20;/h11-13,15,20,27H,5-10,14,16H2,1-4H3;1H/t20-;/m1./s1
SMILES Cl.COc1cc2CCN(CCCNC[C@H]3Cc4cc(OC)c(OC)cc34)C(=O)Cc2cc1OC
IUPAC Name 3-[3-[[(7S)-3,4-dimethoxy-7-bicyclo[4.2.0]octa-1,3,5-trienyl]methylamino]propyl]-7,8-dimethoxy-2,5-dihydro-1H-3-benzazepin-4-one;hydrochloride
Molecular Weight (g/mol) 491.02
Formula Weight 490.2234
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.