missing translation for 'onlineSavingsMsg'
Learn More

N-Deethylcyanazine amide, TRC

Product Code. 30381304
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381304 100 mg 100mg
30372361 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381304 Supplier Toronto Research Chemicals Supplier No. TRCD953505100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36556-77-1
Molecular Formula C7H11ClN6O
Molecular Weight (g/mol) 230.65
Synonym 2-[(4-amino-6-chloro-1,3,5-triazin-2-yl)amino]-2-methyl-Propanamide, 2-[(4-Amino-6-chloro-1,3,5-triazin-2-yl)amino]-2-methylpropanamide, 2-Chloro-4-(1-carbamoyl-1-methylethylamino)-6-amino-1,3,5-triazine, 2-Chloro-4-(1-carbamoyl-1-methylethylamino)-6-aminotriazine
IUPAC Name 2-[(4-amino-6-chloro-1,3,5-triazin-2-yl)amino]-2-methylpropanamide
SMILES CC(C)(Nc1nc(N)nc(Cl)n1)C(=O)N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H11ClN6O/c1-7(2,3(9)15)14-6-12-4(8)11-5(10)13-6/h1-2H3,(H2,9,15)(H3,10,11,12,13,14)
Formula Weight 230.0683
Chemical Name or Material N-Deethylcyanazine amide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.