missing translation for 'onlineSavingsMsg'
Learn More

n-Decyltrimethoxysilane, 97%

Product Code. 11343866
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343866 100 g 100g
11333866 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343866 Supplier Thermo Scientific Alfa Aesar Supplier No. L16966.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5575-48-4 | C13H30O3Si | 262.47 g/mol

n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5575-48-4
Molecular Formula C13H30O3Si
Molecular Weight (g/mol) 262.47
MDL Number MFCD00069135
InChI Key KQAHMVLQCSALSX-UHFFFAOYSA-N
Synonym n-decyltrimethoxysilane, decyltrimethoxysilan, trimethoxydecylsilane, decyl trimethoxy silane, silane,decyltrimethoxy, decyl-trimethoxysilane
PubChem CID 2758023
IUPAC Name decyl(trimethoxy)silane
SMILES CCCCCCCCCC[Si](OC)(OC)OC

Specifications

Boiling Point 102°C to 106°C (3.5mmHg)
Melting Point -37°C
Quantity 100 g
Beilstein 8625082
Flash Point 117°C (242°F)
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 262.47
Percent Purity 98%
Refractive Index 1.422
Sensitivity Moisture sensitive
Density 0.9
Chemical Name or Material n-Decyltrimethoxysilane
Show More Show Less
compliance-icons
Product Identifier
  • n-Decyltrimethoxysilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: decyl Trimethoxysilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.